Läs mer
Chloride (from NH4Cl) Standard: Cl- @ 10000 μg/mL in H2O, VHG™
Produktkod. 17978934
Change view
Kvantitet:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17978934 100 ml
17979224 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17978934 Leverantör VHG Labs Leverantörsnummer VHGI1PACL100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Chloride (from NH4Cl) Standard: Cl- @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloride
CAS 16887-00-6
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnamn Chloride
Frakt skick Room Temperature
Molekylformel Cl-
Synonym Cl−
LEDER [Cl-]
IUPAC-namn Chloride
Lösningsmedel eller matris Water
Molekylvikt (g/mol) 35.45
Kvantitet 100 ml
Formel vikt 35.45 g/mol