Läs mer
Chlorin E6, TRC
Produktkod. 30360207
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360207 500 mg
30358024 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360207 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC380285500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19660-77-6
Molekylformel C34H36N4O6
Molekylvikt (g/mol) 596.67
Synonym (7S,8S)-3-Carboxy-5-(carboxymethyl)-13-ethenyl-18-ethyl-7,8-dihydro-2,8,12,17-tetramethyl-21H,23H-porphine-7-propanoic Acid, 18-Carboxy-20-(carboxymethyl)-13-ethyl-2beta,3-dihydro-3beta,7,12,17-tetramethyl-8-vinyl-2-porphinepropionic Acid, (2S-trans)-18-Carboxy-20-(carboxymethyl)-8-ethenyl-13-ethyl-2,3-dihydro-3,7,12,17-tetramethyl-21H,23H-porphine-2-propanoic Acid, Chlorine E6, Chlorin A6, Chlorin E6, Chlorine A6, Chlorophyllin, Photochlorine, Photolon, Phytochlorin, Phytochlorin E6, Phytochlorine E6
IUPAC-namn (17S,18S)-18-(2-carboxyethyl)-20-(carboxymethyl)-12-ethenyl-7-ethyl-3,8,13,17-tetramethyl-17,18,22,23-tetrahydroporphyrin-2-carboxylic acid
LEDER CCc1c(C)c2cc3[nH]c(cc4nc([C@@H](CCC(=O)O)[C@@H]4C)c(CC(=O)O)c5[nH]c(cc1n2)C(=C5C(=O)O)C)c(C)c3C=C
Specifikationer
Kemiskt namn eller material Chlorin E6
Kvantitet 500 mg
InChI formel InChI=1S/C34H36N4O6/c1-7-19-15(3)23-12-25-17(5)21(9-10-29(39)40)32(37-25)22(11-30(41)42)33-31(34(43)44)18(6)26(38-33)14-28-20(8-2)16(4)24(36-28)13-27(19)35-23/h7,12-14,17,21,35-36H,1,8-11H2,2-6H3,(H,39,40)(H,41,42)(H,43,44)/t17-,21-/m0/s1
Formel vikt 596.2635