Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 250420
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11783177
  • 1225.00 SEK / Pack of 150
Estimated Shipment: 17-06-2024
to see stock.
Specifications
DIN 38408 G4, EN ISO 7393
Disinfectants, wastewater, drinking water
Free and total Chlorine
0.03 to 6.00 Cl2, free and total
Cell Test Kit
5 mL
PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Photoflex™ meter
150 Pack
DPD
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Chlorine Cell Test (free and total chlorine) Method: photometric, DPD 0.03 - 6.00 mg/l Cl
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : boric acid, Ethylenedinitrilotetraaceticacid,Tetrasodiumsalt