Läs mer
Chlormidazole Hydrochloride, TRC
Produktkod. 30363543
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30363543 2.5 g
1 options
Produktkod. 30363543 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3440002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chlormidazole Hydrochloride
CAS 74298-63-8
Molekylformel C15H14Cl2N2
Kvantitet 2.5 g
Synonym 1-[(4-Chlorophenyl)methyl]-2-methyl-1H-benzimidazole Hydrochloride, 1-[(4-Chlorophenyl)methyl]-2-methyl-1H-benzimidazole Monohydrochloride, 1-(p-Chlorobenzyl)-2-methyl-benzimidazole Hydrochloride, Diamyceline, Futrican, H 115
InChI formel InChI=1S/C15H13ClN2.ClH/c1-11-17-14-4-2-3-5-15(14)18(11)10-12-6-8-13(16)9-7-12;/h2-9H,10H2,1H3;1H
LEDER CCNCCCNCc1c(O)c2c(cccc2)nc1
IUPAC-namn 1-[(4-chlorophenyl)methyl]-2-methylbenzimidazole;hydrochloride
Molekylvikt (g/mol) 293.19
Formel vikt 292.0534