Läs mer
Chloro(triethylphosphine)gold, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Chloro(triethylphosphine)gold |
| CAS | 15529-90-5 |
| Molekylformel | C6H15AuClP |
| Kvantitet | 100 mg |
| Synonym | (Triethylphosphine)gold chloride, NSC 313981, SKF 36914, Triethylphosphine gold(I) chloride, Triethylphosphine-gold chloride complex, Triethylphosphineaurous chloride |
| InChI formel | InChI=1S/C6H15P.Au.ClH/c1-4-7(5-2)6-3;;/h4-6H2,1-3H3;;1H/q;+1;/p-1 |
| LEDER | [Cl-].[Au+].CCP(CC)CC |
| IUPAC-namn | gold(1+);triethylphosphane;chloride |
| Molekylvikt (g/mol) | 350.58 |
| Formel vikt | 350.0265 |