missing translation for 'onlineSavingsMsg'
Learn More

Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min, Thermo Scientific Chemicals

Catalog Number p-4824672
Click to view available options
Quantity:
0.5 g
1 g
5 g
This item is not returnable. View return policy
14243-64-2
C18H15AuClP
494.71
MFCD00009588
IFPWCRBNZXUWGC-UHFFFAOYSA-M
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
10874691
chlorogold;triphenylphosphane
Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
14243-64-2
C18H15AuClP
494.71
MFCD00009588
IFPWCRBNZXUWGC-UHFFFAOYSA-M
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
10874691
chlorogold;triphenylphosphane
Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Chloro(triphenylphosphine)gold(I) is a common reagent in gold chemistry. It catalyzes certain rearrangement reactions in organic synthesis. It is also used to prepare 1-methylthyminato-N3-triphenylphosphinegold(I) by reacting with 1-methylthymine. It acts as a precursor for the synthesis of gold(I) and gold(III) organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Assay Percent Range 99.99% (Metal basis, Au 39.3% min.)
Odor Odorless
Quantity 0.5 g
Solubility Information Soluble in methylene chloride,acetonitrile,benzene and acetone. Insoluble in water and ethanol.
Formula Weight 494.71
Physical Form Powder
Chemical Name or Material Chloro(triphenylphosphine)gold(I)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'