missing translation for 'onlineSavingsMsg'
Learn More

α-chlorocinnamaldehyde, +90.5%, ACROS Organics™

Brand:  Acros Organics 393531000

Additional Details : CAS Number : 18365-42-9 Weight : 0.15000kg


Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 10015353

  • 1721.25 SEK / 100g


Estimated Shipment date
from Supplier 01-10-2019
Add to basket


Specifications

Chemical Identifiers

18365-42-9
166.61
SARRRAKOHPKFBW-TWGQIWQCSA-N
5899053
C1=CC=C(C=C1)C=C(C=O)Cl
C9H7ClO
MFCD00077440
alpha-chlorocinnamaldehyde, 2-chloro-3-phenylacrylaldehyde, 2-propenal, 2-chloro-3-phenyl, 2-chloro-3-phenyl-2-propenal, cinnamaldehyde, alpha-chloro, .alpha.-chlorocinnamaldehyde, 2z-2-chloro-3-phenylprop-2-enal, z-2-chloro-3-phenyl-2-propenal, a-chlorocinnamaldehyde, cinnamaldehyde, alpha-chloro-, z
(Z)-2-chloro-3-phenylprop-2-enal

Specifications

100g
>110°C
Glass bottle
107°C to 108°C (3.0mmHg)
20°C to 22°C
90.0
90+%
C6H5CH=C(Cl)CHO
alpha-chlorocinnamaldehyde, 2-chloro-3-phenylacrylaldehyde, 2-propenal, 2-chloro-3-phenyl, 2-chloro-3-phenyl-2-propenal, cinnamaldehyde, alpha-chloro, .alpha.-chlorocinnamaldehyde, 2z-2-chloro-3-phenylprop-2-enal, z-2-chloro-3-phenyl-2-propenal, a-chlorocinnamaldehyde, cinnamaldehyde, alpha-chloro-, z
C1=CC=C(C=C1)C=C(C=O)Cl
166.61
166.61
≥90.0%
α-chlorocinnamaldehyde
Authentic
1.6390 to 1.641
Brown
18365-42-9
100.0
C9H7ClO
MFCD00077440
SARRRAKOHPKFBW-TWGQIWQCSA-N
(Z)-2-chloro-3-phenylprop-2-enal
5899053
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • a-Chlorocinnamaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents