Learn More
Chlorocycloheptane, 96%, BLD Pharm™
Product Code. 30948335
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948335 5 g 5g
30939678 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30948335 Supplier BLD Pharm Supplier No. BD1226635g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 2453-46-5
Molecular Formula C7H13Cl
MDL Number MFCD00046501
Quantity 5 g
SMILES ClC1CCCCCC1
Molecular Weight (g/mol) 132.63
Percent Purity 96%
Chemical Name or Material Chlorocycloheptane