missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl-n-propylsilane, 97%, Thermo Scientific™

Catalog Number 11309295
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
17477-29-1
C5H13ClSi
136.694
MFCD00039301
HXVPUKPVLPTVCQ-UHFFFAOYSA-N
propyldimethylchlorosilane, chlorodimethylpropylsilane, n-propyldimethylchlorosilane, silane, chlorodimethylpropyl, dimethylpropylchlorosilane, chlorodimethyl propyl silane, chlorodimethyl-n-propylsilane
87130
chloro-dimethyl-propylsilane
CCC[Si](C)(C)Cl
This item is not returnable. View return policy
17477-29-1
C5H13ClSi
136.694
MFCD00039301
HXVPUKPVLPTVCQ-UHFFFAOYSA-N
propyldimethylchlorosilane, chlorodimethylpropylsilane, n-propyldimethylchlorosilane, silane, chlorodimethylpropyl, dimethylpropylchlorosilane, chlorodimethyl propyl silane, chlorodimethyl-n-propylsilane
87130
chloro-dimethyl-propylsilane
CCC[Si](C)(C)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 113°C to 114°C
Quantity 5 g
UN Number UN2985
Beilstein 1732132
Flash Point 8°C (46°F)
Formula Weight 136.7
Percent Purity 97%
Refractive Index 1.414
Sensitivity Moisture sensitive
Density 0.873
Chemical Name or Material Chlorodimethyl-n-propylsilane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'