Läs mer
Chloroform Water - British Pharmacopoeia, Fisherbrand™
Produktkod. 30695075 Handla allt Fisher Chemical Produkter
Change view
Kvantitet:
1000 mL
Produktkod. Kvantitet
30695075 1000 mL
1 options
Produktkod. 30695075 Leverantör Fisher Chemical Leverantörsnummer BPR 079
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Pharmacopoeia reagents for testing and monographs, produced under a GMP-compliant ISO 9001 quality management system.
    This Pharmacopoeia reagent is manufactured :
  • Using pharmacopoeia reagent quality components
  • Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
  • Supplied with a Certificate of Analysis for Performance Test
  • Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034
Specifikationer
Kvantitet 1000 mL