Läs mer
Chloromethyl Diethylcarbamate, TRC
Produktkod. 30360157
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30360157 100 mg
1 options
Produktkod. 30360157 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC369125100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloromethyl Diethylcarbamate
CAS 133217-92-2
Molekylformel C6H12ClNO2
Kvantitet 100 mg
Synonym Diethyl-carbamic Acid Chloromethyl Ester
InChI formel InChI=1S/C6H12ClNO2/c1-3-8(4-2)6(9)10-5-7/h3-5H2,1-2H3
LEDER CCN(CC)C(=O)OCCl
IUPAC-namn chloromethyl N,N-diethylcarbamate
Molekylvikt (g/mol) 165.62
Formel vikt 165.06