Läs mer
Chloromethyl Dimethylcarbamate, TRC
Produktkod. 30364345
Change view
Kvantitet:
2.5 g
Förpackningsstorlek:
2.5g
Produktkod. Kvantitet unitSize
30364345 2.5 g 2.5g
1 options
Produktkod. 30364345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5878882.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloromethyl Dimethylcarbamate
CAS 49622-08-4
Molekylformel C4H8ClNO2
Kvantitet 2.5 g
Synonym Chloromethyl N,N-Dimethylcarbamate
InChI formel InChI=1S/C4H8ClNO2/c1-6(2)4(7)8-3-5/h3H2,1-2H3
LEDER CN(C)C(=O)OCCl
IUPAC-namn chloromethyl N,N-dimethylcarbamate
Molekylvikt (g/mol) 137.56
Formel vikt 137.0244