Läs mer
Chloromethyl Dimethylcarbamate, TRC
Produktkod. 30363835
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30363835 250 mg
1 options
Produktkod. 30363835 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC587888250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloromethyl Dimethylcarbamate
CAS 49622-08-4
Molekylformel C4H8ClNO2
Kvantitet 250 mg
Synonym Chloromethyl N,N-Dimethylcarbamate
InChI formel InChI=1S/C4H8ClNO2/c1-6(2)4(7)8-3-5/h3H2,1-2H3
LEDER CN(C)C(=O)OCCl
IUPAC-namn chloromethyl N,N-dimethylcarbamate
Molekylvikt (g/mol) 137.56
Formel vikt 137.0244