Läs mer
Chloromethyl Isobutyrate, TRC
Produktkod. 30359803
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30359803 1 g -
30356782 250 mg -
30352651 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359803 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3819501G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61644-18-6
Molekylformel C5H9O2Cl
Molekylvikt (g/mol) 136.57
Synonym (Isobutyryloxy)methyl chloride, Isobutyric acid chloromethyl ester, Chloromethyl 2-methylpropionate, Chloromethyl 2-methylpropanoate
IUPAC-namn chloromethyl 2-methylpropanoate
LEDER CC(C)C(=O)OCCl
Specifikationer
Kemiskt namn eller material Chloromethyl Isobutyrate
Kvantitet 1 g
InChI formel InChI=1S/C5H9ClO2/c1-4(2)5(7)8-3-6/h4H,3H2,1-2H3
Formel vikt 136.0291