Läs mer
Chloromethyl Laurate, TRC
Produktkod. 30362840
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30362840 1 g
1 options
Produktkod. 30362840 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3710051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloromethyl Laurate
CAS 61413-67-0
Molekylformel C13 H25 Cl O2
Kvantitet 1 g
Synonym Dodecanoic Acid Chloromethyl Ester, Chloromethyl Dodecanoate, Dodecanoyloxymethyl Chloride
InChI formel InChI=1S/C13H25ClO2/c1-2-3-4-5-6-7-8-9-10-11-13(15)16-12-14/h2-12H2,1H3
LEDER CCCCCCCCCCCC(=O)OCCl
IUPAC-namn chloromethyl dodecanoate
Molekylvikt (g/mol) 248.79
Formel vikt 248.1543