Läs mer
Chloromethyl Methyl Carbonate, TRC
Produktkod. 30358160
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30358160 5 g
1 options
Produktkod. 30358160 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3693055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloromethyl Methyl Carbonate
CAS 40510-81-4
Molekylformel C3 H5 Cl O3
Kvantitet 5 g
Synonym Tenofovir Impurity 52
InChI formel InChI=1S/C3H5ClO3/c1-6-3(5)7-2-4/h2H2,1H3
LEDER COC(=O)OCCl
IUPAC-namn chloromethyl methyl carbonate
Molekylvikt (g/mol) 124.52
Formel vikt 123.9927