Läs mer
Chloromethyl Propyl Carbonate, TRC
Produktkod. 30360412
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360412 500 mg
30366496 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360412 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC369225500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35273-90-6
Molekylformel C5H9ClO3
Molekylvikt (g/mol) 152.58
Synonym Chloromethyl Carbonic Acid Propyl Ester
IUPAC-namn chloromethyl propyl carbonate
LEDER CCCOC(=O)OCCl
Specifikationer
Kemiskt namn eller material Chloromethyl Propyl Carbonate
Kvantitet 500 mg
InChI formel InChI=1S/C5H9ClO3/c1-2-3-8-5(7)9-4-6/h2-4H2,1H3
Formel vikt 152.024