Läs mer
Chlorophosphonazo III (Technical Grade), TRC
Produktkod. 30364017
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364017 1 g
30353306 500 mg
30351584 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364017 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3753851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1914-99-4
Molekylformel C22H16Cl2N4O14P2S2
Molekylvikt (g/mol) 757.36
Synonym 3,6-bis[(4-Chloro-2-phosphonophenyl)azo]-4,5-dihydroxy-2,7-naphthalenedisulfonic Acid, 3,6-bis[2-(4-Chloro-2-phosphonophenyl)diazenyl]-4,5-dihydroxy-2,7-naphthalenedisulfonic Acid, Chlorphosphonazo III, Dichlorophosphonazo III
IUPAC-namn 3,6-bis[(E)-(4-chloro-2-phosphonophenyl)diazenyl]-4,5-dihydroxynaphthalene-2,7-disulfonic acid
LEDER Oc1c(N=Nc2ccc(Cl)cc2P(=O)(O)O)c(cc3cc(c(N=Nc4ccc(Cl)cc4P(=O)(O)O)c(O)c13)S(=O)(=O)O)S(=O)(=O)O
Specifikationer
Kemiskt namn eller material Chlorophosphonazo III (Technical Grade)
Kvantitet 1 g
InChI formel InChI=1S/C22H16Cl2N4O14P2S2/c23-10-1-3-12(14(7-10)43(31,32)33)25-27-19-16(45(37,38)39)5-9-6-17(46(40,41)42)20(22(30)18(9)21(19)29)28-26-13-4-2-11(24)8-15(13)44(34,35)36/h1-8,29-30H,(H2,31,32,33)(H2,34,35,36)(H,37,38,39)(H,40,41,42)
Formel vikt 755.8957