Läs mer
Chloroprocaine Hydrochloride, LoGiCal
Produktkod. 30460009
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30460009 5 mg
1 options
Produktkod. 30460009 Leverantör LoGiCal Leverantörsnummer LGCFOR1994.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloroprocaine Hydrochloride
CAS 3858-89-7
Analyt- eller komponentnamn Chloroprocaine Hydrochloride
Frakt skick Room Temperature
Molekylformel C13 H19 Cl N2 O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Benzoic acid, 4-amino-2-chloro-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-amino-2-chloro-, 2-(diethylamino)ethyl ester, monohydrochloride (8CI,9CI), 2-(Diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride, 2-Chloro-4-aminobenzoic acid diethylaminoethyl ester hydrochloride, 2-Chloroprocaine hydrochloride, Chloroprocaine hydrochloride, Chlorprocaine hydrochloride, Nesacaine, Nesacaine hydrochloride, Chloroprocaine Hydrochloride
InChI formel InChI=1S/C13H19ClN2O2.ClH/c1-3-16(4-2)7-8-18-13(17)11-6-5-10(15)9-12(11)14;/h5-6,9H,3-4,7-8,15H2,1-2H3;1H
Lösningstyp Neat
LEDER Cl.CCN(CC)CCOC(=O)c1ccc(N)cc1Cl
IUPAC-namn 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 307.22
Kvantitet 5 mg
Formel vikt 306.0902 g/mol