Läs mer
Chloroquine Sulfate, TRC
Produktkod. 17913915
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17913915 25 mg
17940535 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17913915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38031525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6823-83-2
Molekylformel C18H26ClN3.H2O.H2O4S
Molekylvikt (g/mol) 435.97
Synonym Chloroquine sulfate, N4-(7-chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine sulfate
IUPAC-namn 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;sulfuric acid;hydrate
LEDER O.CCN(CC)CCCC(C)Nc1ccnc2cc(Cl)ccc12.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Chloroquine sulfate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C18H26ClN3.H2O4S.H2O/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18;1-5(2,3)4;/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21);(H2,1,2,3,4);1H2
Formel vikt 435.1595
Procent renhet >95%