missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroquine, TRC
Specifications
Specifications
| Kemiskt namn eller material | Chloroquine |
| CAS | 54-05-7 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C18 H26 Cl N3 |
| Kvantitet | 10 mg |
| Synonym | N4-(7-Chloro-4-quinolinyl)-N1,N1-diethyl-1,4-pentanediamine, (±)-Chloroquine, 7-Chloro-4-[[4-(diethylamino)-1-methylbutyl]amino]quinoline, Aralen, Artrichin, Bipiquin, Capquin, Chloraquine, Chlorochin, Chlorolex, loquin, E-Vivax, Hcqs, Maliago, NSC 187208, Nivaquinp, Premoquine, RP 3377, Reumachlor, Ronaquine, Rosq, ST 121, ST 121 (Pharmaceutical), Sequin |
| InChI formel | InChI=1S/C18H26ClN3/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21) |
| LEDER | CCN(CC)CCCC(C)Nc1ccnc2cc(Cl)ccc12 |
| IUPAC-namn | 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine |
| Molekylvikt (g/mol) | 319.87 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?