Läs mer
Chloroquine, TRC
Produktkod. 30353825
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30353825 50 mg
1 options
Produktkod. 30353825 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38031850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chloroquine
CAS 54-05-7
Anteckningar om renhetsgrad HPLC
Molekylformel C18 H26 Cl N3
Kvantitet 50 mg
Synonym N4-(7-Chloro-4-quinolinyl)-N1,N1-diethyl-1,4-pentanediamine, (±)-Chloroquine, 7-Chloro-4-[[4-(diethylamino)-1-methylbutyl]amino]quinoline, Aralen, Artrichin, Bipiquin, Capquin, Chloraquine, Chlorochin, Chlorolex, loquin, E-Vivax, Hcqs, Maliago, NSC 187208, Nivaquinp, Premoquine, RP 3377, Reumachlor, Ronaquine, Rosq, ST 121, ST 121 (Pharmaceutical), Sequin
InChI formel InChI=1S/C18H26ClN3/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21)
LEDER CCN(CC)CCCC(C)Nc1ccnc2cc(Cl)ccc12
IUPAC-namn 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine
Molekylvikt (g/mol) 319.87
Formel vikt 319.1815