missing translation for 'onlineSavingsMsg'
Learn More

chlorotripropylsilane, 98%, ACROS Organics™

Brand:  Acros Organics 313930050

Additional Details : CAS Number : 995-25-5 Weight : 0.05500kg Transport : UN number : 2987 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11955841

  • 600.00 SEK / 5g

In stock, ready to ship: 4

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

995-25-5
192.81
tri-n-propylchlorosilane, tripropylchlorosilane, silane, chlorotripropyl, chlorotri-n-propylsilane, chloro tripropyl silane, chloro-tri-n-propylsilane, chlorotripropylsilane, chloro tri-n-propyl silane
chloro(tripropyl)silane
C9H21ClSi
ACTAPAGNZPZLEF-UHFFFAOYSA-N
70445
CCC[Si](CCC)(CCC)Cl

Specifications

chlorotripropylsilane
76°C
Glass bottle
0.882
Colorless
995-25-5
C9H21ClSi
ACTAPAGNZPZLEF-UHFFFAOYSA-N
chloro(tripropyl)silane
70445
Liquid
0.8820g/mL
Authentic
1.3990 to 1.4410 (20°C, 589nm)
199.0°C to 200.0°C
5g
97.5% min. (GC)
tri-n-propylchlorosilane, tripropylchlorosilane, silane, chlorotripropyl, chlorotri-n-propylsilane, chloro tripropyl silane, chloro-tri-n-propylsilane, chlorotripropylsilane, chloro tri-n-propyl silane
CCC[Si](CCC)(CCC)Cl
192.81
192.81
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chlorotripropylsilane

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents
Certificates