Läs mer
Chlorphentermine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Chlorphentermine Hydrochloride |
| CAS | 151-06-4 |
| Molekylformel | C10 H14 Cl N . Cl H |
| Kvantitet | 500 mg |
| Synonym | Chlorphentermine Hydrochloride, Benzeneethanamine, 4-chloro-alpha,alpha-dimethyl-, hydrochloride (1:1), Benzeneethanamine, 4-chloro-alpha,alpha-dimethyl-, hydrochloride (9CI), Phenethylamine, p-chloro-alpha,alpha-dimethyl-, hydrochloride (6CI,8CI), 1-(p-Chlorophenyl)-2-methyl-2-aminopropane hydrochloride, 4-Chloro-alpha,alha-dimethylphenethylamine hydrochloride, Apsedon, Avicol, Avicol (pharmaceutical), Avipron, Chlorophentermine hydrochloride, Chlorphentermine hydrochloride, Chlorphenterminium chloride, Clomina, Dezopimon, Lucofen, Lucofene, Lukofen hydrochloride, NSC 76098, Nilgana, Pre-Sate, Presate hydrochloride, S 62, S 62-2, W 2426, WX 2426, p-Chloro-alpha,alpha-dimethylphenethylamine hydrochloride, p-Chlorphentermine hydrochloride, 4-Chloro-alpha,alpha-dimethylphenethylamine hydrochloride |
| InChI formel | InChI=1S/C10H14ClN.ClH/c1-10(2,12)7-8-3-5-9(11)6-4-8;/h3-6H,7,12H2,1-2H3;1H |
| LEDER | Cl.CC(C)(N)Cc1ccc(Cl)cc1 |
| IUPAC-namn | 1-(4-chlorophenyl)-2-methylpropan-2-amine;hydrochloride |
| Molekylvikt (g/mol) | 220.14 |
| Formel vikt | 219.0582 |