Läs mer
Chlorpromazine N-Oxide Maleic Acid Salt, TRC
Produktkod. 30134422
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134422 100 mg
30008672 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134422 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC424745100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18683-80-2
Molekylformel C21H23ClN2O5S
Molekylvikt (g/mol) 450.94
Synonym 2-Chloro-N,N-dimethyl-10H-phenothiazine-10-propanamine N-oxide, (2Z)-2-butenedioate, 2-Chloro-10-[3-(dimethylamino)propyl]-phenothiazine N-oxide, maleate, Chlorpromazine N-Oxide Maleate
IUPAC-namn (Z)-but-2-enedioic acid;3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine oxide
LEDER C[N+](C)([O-])CCCN1c2ccccc2Sc3ccc(Cl)cc13.OC(=O)\C=C/C(=O)O
Specifikationer
Kemiskt namn eller material Chlorpromazine N-Oxide Maleic Acid Salt
Kvantitet 100 mg
InChI formel InChI=1S/C17H19ClN2OS.C4H4O4/c1-20(2,21)11-5-10-19-14-6-3-4-7-16(14)22-17-9-8-13(18)12-15(17)19;5-3(6)1-2-4(7)8/h3-4,6-9,12H,5,10-11H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vikt 450.1016