Läs mer
Chlorpromazine N-Oxide, TRC
Produktkod. 30358230
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30358230 10 mg
1 options
Produktkod. 30358230 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC59643010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chlorpromazine N-Oxide
CAS 1672-76-0
Molekylformel C17 H19 Cl N2 O S
Kvantitet 10 mg
Synonym 2-Chloro-N,N-dimethyl-10H-phenothiazine-10-propanamine N-oxide, Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]-, N-oxide (7CI,8CI), Ba 2836, Chlorpromazin N-oxide, Chlorpromazine N-oxide, Chlorpromazine oxide
InChI formel InChI=1S/C17H19ClN2OS/c1-20(2,21)11-5-10-19-14-6-3-4-7-16(14)22-17-9-8-13(18)12-15(17)19/h3-4,6-9,12H,5,10-11H2,1-2H3
LEDER C[N+](C)([O-])CCCN1c2ccccc2Sc3ccc(Cl)cc13
IUPAC-namn 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine oxide
Molekylvikt (g/mol) 334.86
Formel vikt 334.0907