Läs mer
Chlorsulfaquinoxaline, TRC
Produktkod. 30366539
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366539 5 mg
30364005 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366539 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4260005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chlorsulfaquinoxaline
CAS 97919-22-7
Molekylformel C14 H11 Cl N4 O2 S
Kvantitet 5 mg
Synonym 4-Amino-N-(5-chloro-2-quinoxalinyl)benzenesulfonamide, CQS, NSC 339004
InChI formel InChI=1S/C14H11ClN4O2S/c15-11-2-1-3-12-14(11)17-8-13(18-12)19-22(20,21)10-6-4-9(16)5-7-10/h1-8H,16H2,(H,18,19)
LEDER Nc1ccc(cc1)S(=O)(=O)Nc2cnc3c(Cl)cccc3n2
IUPAC-namn 4-amino-N-(5-chloroquinoxalin-2-yl)benzenesulfonamide
Molekylvikt (g/mol) 334.78
Formel vikt 334.0291