Läs mer
Chlorthalidone Acid Ethyl Ester, TRC
Produktkod. 30359881
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359881 10 mg
30351044 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359881 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC42750510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 92874-73-2
Molekylformel C16H14ClNO5S
Molekylvikt (g/mol) 367.8
Synonym 2-[3-(Aminosulfonyl)-4-chlorobenzoyl]-benzoic Acid Ethyl Ester, o-(4-Chloro-3-sulfamoylbenzoyl)-benzoic Acid Ethyl Ester
IUPAC-namn ethyl 2-(4-chloro-3-sulfamoylbenzoyl)benzoate
LEDER CCOC(=O)c1ccccc1C(=O)c2ccc(Cl)c(c2)S(=O)(=O)N
Specifikationer
Kemiskt namn eller material Chlorthalidone Acid Ethyl Ester
Kvantitet 10 mg
InChI formel InChI=1S/C16H14ClNO5S/c1-2-23-16(20)12-6-4-3-5-11(12)15(19)10-7-8-13(17)14(9-10)24(18,21)22/h3-9H,2H2,1H3,(H2,18,21,22)
Formel vikt 367.0281