Läs mer
Chlorthion, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Chlorthion |
| CAS | 500-28-7 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C8 H9 Cl N O5 P S |
| Kvantitet | 1 g |
| Synonym | Phosphorothioic acid, O-(3-chloro-4-nitrophenyl) O,O-di-Me ester (6CI), BAY 22190, Chlorothion, Chlorthion, Chlorthion methyl, Compound 22/190, Methylchlorothion, NSC 8927, O,O-Dimethyl O-(3-chloro-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(3-chloro-4-nitrophenyl)thionophosphate, O,O-Dimethyl O-3-chloro-4-nitrophenyl phosphorothioate, O-(3-Chloro-4-nitrophenyl) O,O-dimethyl phosphorothioate, OMS 217 |
| InChI formel | InChI=1S/C8H9ClNO5PS/c1-13-16(17,14-2)15-6-3-4-8(10(11)12)7(9)5-6/h3-5H,1-2H3 |
| LEDER | COP(=S)(OC)Oc1ccc(c(Cl)c1)[N+](=O)[O-] |
| IUPAC-namn | (3-chloro-4-nitrophenoxy)-dimethoxy-sulfanylidene-λ^{5}-phosphane |
| Molekylvikt (g/mol) | 297.65 |
| Visa mer |