Learn More
Chlorzoxazone, TRC
Product Code. 30132456
Change view
Click to view available options
Quantity:
10 mg
1 g
10 g
Unit Size:
10g
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132456 10 g 10g
30135302 1 g 1g
17974805 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132456 Supplier Toronto Research Chemicals Supplier No. TRCC42870010G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-25-0
Molecular Formula C7 H4 Cl N O2
Molecular Weight (g/mol) 169.57
Synonym Chlorzoxazone, 2-Benzoxazolinone, 5-chloro- (6CI,7CI,8CI), 2-Hydroxy-5-chlorobenzoxazole, 5-Chloro-1,3-benzoxazol-2(3H)-one, 5-Chloro-2(3H)-benzoxazolone, 5-Chloro-2-benzoxazolinone, 5-Chloro-2-benzoxazolol, 5-Chloro-2-benzoxazolone, 5-Chloro-2-hydroxybenzoxazole, 5-Chloro-2-oxo-3H-benzoxazole, 5-Chloro-3H-benzoxazol-2-one, 5-Chlorobenzoxazolidone, 5-Chlorobenzoxazolone, Biomioran, Chloroxazone, Chlorzoxazon, Chlorzoxazone, Cipzox, Escoflex, Mioran, Miotran, Myoflexin, Myoflexine, NSC 26189, Neoflex, Paraflex, Parafon Forte DSC, Parfon-forte, Pathorysin, Solaxin, 2(3H)-Benzoxazolone, 5-chloro-
IUPAC Name 5-chloro-3H-1,3-benzoxazol-2-one
SMILES Clc1ccc2OC(=O)Nc2c1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H4ClNO2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)
Formula Weight 168.9931
Percent Purity >95%
Chemical Name or Material Chlorzoxazone
Grade HPLC