missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CHML Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556213
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: YASQDMEDNV EEIGALQKNP SLGVSNTFTE VLDSALPEES QLSYFNSDEM PAKHTQKSDT EISLE Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 39%.
Binds unprenylated Rab proteins, presents it to the catalytic component B, and remains bound to it after the geranylgeranyl transfer reaction. The component A may be regenerated by transferring its prenylated Rab to a protein acceptor. Less effective than REP-1 in supporting prenylation of Rab3 family.
Specifikationer
| CHML | |
| Polyclonal | |
| Unconjugated | |
| CHML | |
| AI450814; CHM like, Rab escort protein 2; Chml; choroideraemia-like protein; choroideremia-like; choroideremia-like (Rab escort protein 2); choroideremia-like protein; E030003F13Rik; rab escort protein 2; rab escort protein-2; rab proteins geranylgeranyltransferase component A 2; REP2; REP-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26374 | |
| CHML | |
| Recombinant protein corresponding to Human CHML. Recombinant protein control fragment (Product #RP-94561). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering