missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14639-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
CHMP1B belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).Specifications
CHMP1B | |
Polyclonal | |
Unconjugated | |
CHMP1B | |
C10orf2, C18 ORF2, C18orf2, CHMP1.5, CHMP1B, chromatin modifying protein 1B, hVps46 2, Vps46 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57132, 67064, 689364 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7LBR1 | |
CHMP1B | |
CHMP1B Fusion Protein Ag6222 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |