Läs mer
DiscoverX CHO-K1 GPR17 Gq Cell Line
Produktkod. 17364749
Encompass
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2 injektionsflaskor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17364749 2 injektionsflaskor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 17364749 Leverantör DiscoverX Leverantörsnummer 951022C2
 Begränsad artikel
Beräknad leverans: 07-10-2026
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
The CHO-K1 GPR17 Gq Cell Line measures GPR17 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
The GPR17 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MDL 29,951) based activation of GPR17 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPR17 , a naturally Gq coupled GPCR. Upon activation of GPR17 , the resulting calcium activity can be fluorescently assayed using the Calcium NoWash PLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifikationer
Analysinträde Calcium Signaling
Celllinje CHO-K1
För användning med (applikation) Cell Lines
Primär agonist MDL 29,951
Kvantitet 2 injektionsflaskor
Målinträde GPR17
Tillträdesnummer NM_005291