Läs mer
Chocolate Brown HT (Technical Grade), TRC
Produktkod. 30353232
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30353232 50 g
1 options
Produktkod. 30353232 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC44000050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Brown HT
CAS 4553-89-3
Molekylformel C27 H18 N4 O9 S2 . 2 Na
Kvantitet 50 g
Synonym 1-Naphthalenesulfonic acid, 4,4'-[[2,4-dihydroxy-5-(hydroxymethyl)-1,3-phenylene]bis(2,1-diazenediyl)]bis-, sodium salt (1:2), 1-Naphthalenesulfonic acid, 4,4'-[[2,4-dihydroxy-5-(hydroxymethyl)-1,3-phenylene]bis(azo)]bis-, disodium salt (9CI), C.I. Food Brown 3 (7CI), C.I. Food Brown 3, disodium salt (8CI), 1538 Brown, Brown HT, C.I. 20285, Certicol Brown N Extra, Certicol Brown RS, Chocolate Brown HT, E 155, Edicol Brown HT, Hexacol Chocolate Brown HT
InChI formel InChI=1S/C27H20N4O9S2.2Na/c32-14-15-13-22(30-28-20-9-11-23(41(35,36)37)18-7-3-1-5-16(18)20)27(34)25(26(15)33)31-29-21-10-12-24(42(38,39)40)19-8-4-2-6-17(19)21;;/h1-13,32-34H,14H2,(H,35,36,37)(H,38,39,40);;/q;2*+1/p-2/b30-28+,31-29+;;
LEDER [Na+].[Na+].OCc1cc(N=Nc2ccc(c3ccccc23)S(=O)(=O)[O-])c(O)c(N=Nc4ccc(c5ccccc45)S(=O)(=O)[O-])c1O
IUPAC-namn disodium;4-[(E)-[2,4-dihydroxy-5-(hydroxymethyl)-3-[(E)-(4-sulfonatonaphthalen-1-yl)diazenyl]phenyl]diazenyl]naphthalene-1-sulfonate
Molekylvikt (g/mol) 652.56
Formel vikt 652.0311