Learn More
Thermo Scientific™ Choice™ PVDF (Hydrophilic) Syringe Filter, 250/Pk.
Use Choice PVDF (hydrophillic) Syringe Filters for general biological filtration, especially when high protein recovery is desired. They do not require pre-wetting.
4299.20 SEK - 4645.22 SEK
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | PVDF (Hydrophilic) |
Sterility | Non-sterile |
For Use With (Application) | HPLC, SPE |
Housing Material | Solvent Resistent, Low-Extractable Polypropylene |
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
17765044
|
Thermo Scientific™
CHO2213-PV-250 |
0.22 μm | 13 mm |
4645.22 SEK
Pack of 250 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
17705054
|
Thermo Scientific™
CHO4513-PV-250 |
0.45 μm | 13 mm |
4645.22 SEK
Pack of 250 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
17735054
|
Thermo Scientific™
CHO4525-PV-250 |
0.45 μm | 25 mm |
4299.20 SEK
Pack of 250 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
Description
Specifications- Membrane: HPLC certified PVDF
- Maximum operating temperature: 100 °C
- Housing: Medical grade, virgin polypropylene
- Autoclave: Sterilize by dry heat at 121 °C for 15 minutes
- HPLC and organic-solvent sample preparation and clean-up
- Protein-based samples with high non-specific binding
- Environmental water samples
- DMF, DMSO, MEK, acetone and most caustic solutions >6N
Specifications
Single-use | |
Non-sterile | |
Solvent Resistent, Low-Extractable Polypropylene | |
No | |
Syringe Filter |
PVDF (Hydrophilic) | |
HPLC, SPE | |
250 Pk. | |
90 psi |