Läs mer
Cholesterol myristate, MedChemExpress
Beskrivning
Cholesterol myristate is a natural steroid present in traditional Chinese medicine. Cholesterol myristate binds to several ion channels such as the nicotinic acetylcholine receptor, GABAA receptor, and the inward-rectifier potassium ion channel.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Cholesterol myristate |
| CAS | 1989-52-2 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C41H72O2 |
| Kvantitet | 250 mg |
| Synonym | Cholesteryl myristate Cholesteryl tetradecanoate |
| Löslighetsinformation | Ethanol : 3.33 mg/mL (5.58 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| LEDER | C[C@@]12[C@]3([H])[C@](CC=C1C[C@@H](OC(CCCCCCCCCCCCC)=O)CC2)([H])[C@@]4([H])[C@@](C)([C@]([C@H](C)CCCC(C)C)([H])CC4)CC3 |
| Molekylvikt (g/mol) | 597.01 |
| Visa mer |