Läs mer
Cholesteryl Acetate Dibromide(Mixture of Diastereomers), TRC
Produktkod. 30365367
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30365367 100 mg
1 options
Produktkod. 30365367 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC432530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cholesteryl Acetate Dibromide (Mixture of Diastereomers)
CAS 514-50-1
Molekylformel C29H48Br2O2
Kvantitet 100 mg
Synonym 3β-Acetoxy-5,6β-dibromo-5α-cholestane, 5,6β-Dibromo-5α-cholestan-3β-ol acetate, 5,6β-Dibromo-5α-cholestan-3β-yl acetate, 5α,6β-Dibromocholesterol acetate, Acebrochol, 5,6β-Dibromo-5α-cholestan-3β-ol Acetate
InChI formel InChI=1S/C29H48Br2O2/c1-18(2)8-7-9-19(3)23-10-11-24-22-16-26(30)29(31)17-21(33-20(4)32)12-15-28(29,6)25(22)13-14-27(23,24)5/h18-19,21-26H,7-17H2,1-6H3/t19-,21+,22+,23-,24+,25+,26-,27-,28-,29+/m1/s1
LEDER [H][C@@]1([C@H](C)CCCC(C)C)[C@@]2(C)CC[C@@]3([H])[C@@]([C@]2([H])CC1)([H])CC(Br)C4(Br)C[C@@H](OC(C)=O)CC[C@@]43C
IUPAC-namn [(3S,5R,6R,8S,9S,10R,13R,14S,17R)-5,6-dibromo-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,3,4,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-yl] acetate
Molekylvikt (g/mol) 588.5
Formel vikt 586.2021