Läs mer
Cholesteryl Arachidonate, TRC
Produktkod. 30363655
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30363655 25 mg
1 options
Produktkod. 30363655 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43258325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cholesteryl Arachidonate
CAS 604-34-2
Molekylformel C47H76O2
Kvantitet 25 mg
Synonym Cholesterol 5Z,8Z,11Z,14Z-eicosatetraenoic Acid Ester, Arachidonic Acid Cholesteryl Ester, (all-Z)-3-[(5Z,8Z,11Z,14Z)-5,8,11,14-eicosatetraenoate] (3beta)-Cholest-5-en-3-ol
InChI formel InChI=1S/C47H76O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-27-45(48)49-40-32-34-46(5)39(36-40)28-29-41-43-31-30-42(38(4)26-24-25-37(2)3)47(43,6)35-33-44(41)46/h11-12,14-15,17-18,20-21,28,37-38,40-44H,7-10,13,16,19,22-27,29-36H2,1-6H3/b12-11-,15-14-,18-17-,21-20-/t38-,40+,41+,42-,43+,44+,46+,47-/m1/s1
LEDER CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C
IUPAC-namn [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Molekylvikt (g/mol) 673.11
Formel vikt 672.5845