Läs mer
Cholesteryl Oleate, TRC
Produktkod. 30363650
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30363650 100 mg
1 options
Produktkod. 30363650 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC432553100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cholesteryl oleate
CAS 303-43-5
Molekylformel C45 H78 O2
Kvantitet 100 mg
Synonym Cholest-5-en-3-β-yl oleate, Chloesteryl cis-9-octadecanoate, Cholesteryl oleic ester, Oleoylcholesterol
InChI formel InChI=1S/C45H78O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-25-43(46)47-38-30-32-44(5)37(34-38)26-27-39-41-29-28-40(36(4)24-22-23-35(2)3)45(41,6)33-31-42(39)44/h14-15,26,35-36,38-42H,7-13,16-25,27-34H2,1-6H3/b15-14-/t36-,38+,39+,40-,41+,42+,44+,45-/m1/s1
LEDER CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C
IUPAC-namn [(3S,8S,9S,10R,13R,14S,17R)-17-[(1R)-1,5-dimethylhexyl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] (Z)-octadec-9-enoate
Molekylvikt (g/mol) 651.1
Formel vikt 650.6002