Läs mer
Cholic Acid 3-O-Beta-Glucuronide Disodium Salt, TRC
Produktkod. 17962845
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17962845 10 mg
17944435 5 mg
17923365 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17962845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43262510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59274-69-0
Molekylformel C30 H46 O11 . 2 Na
Molekylvikt (g/mol) 628.66
Synonym β-D-Glucopyranosiduronic acid, (3α,5β,7α,12α)-23-carboxy-7,12-dihydroxy-24-norcholan-3-yl, disodium salt (9CI), 24-Norcholane, β-D-glucopyranosiduronic acid deriv., (3α,5β,7α,12α)-23-Carboxy-7,12-dihydroxy-24-norcholan-3-yl β-D-glucopyranosiduronic acid disodium salt, Cholic acid 3-O-glucuronide disodium salt, Cholic acid 3-O-β-glucuronide disodium salt, Cholic acid 3-O-β-D-glucuronide disodium salt, Cholic Acid 3-O-β-Glucuronide Disodium Salt
IUPAC-namn disodium;(2S,3S,4S,5R,6R)-6-[[(3R,5R,7R,8R,9S,10S,12S,13R,14S,17R)-17-[(2R)-4-carboxylatobutan-2-yl]-7,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylate
LEDER [Na+].[Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C)O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material Cholic Acid 3-O-beta-Glucuronide Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C30H48O11.2Na/c1-13(4-7-21(33)34)16-5-6-17-22-18(12-20(32)30(16,17)3)29(2)9-8-15(10-14(29)11-19(22)31)40-28-25(37)23(35)24(36)26(41-28)27(38)39;;/h13-20,22-26,28,31-32,35-37H,4-12H2,1-3H3,(H,33,34)(H,38,39);;/q;2*+1/p-2/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24+,25-,26+,28-,29+,30-;;/m1../s1
Formel vikt 628.2836
Procent renhet >95%