missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline chloride, 99%, MP Biomedicals™
Click to view available options
Quantity:
1 kg
5 kg
Specifications
Color | White |
Density | 1.1g/cm3 at 20°C |
Assay Percent Range | 0.99 |
Quantity | 1 kg |
Formula Weight | 139.63 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Choline chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'