missing translation for 'onlineSavingsMsg'
Learn More

Choline chloride, cell culture reagent, ≥99%, MP Biomedicals™

Catalog Number p-4405720
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
67-48-1
C5H14ClNO
139.62
MFCD00011721
SGMZJAMFUVOLNK-UHFFFAOYSA-M
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
6209
CHEBI:133341
(2-hydroxyethyl)trimethylazanium chloride
[Cl-].C[N+](C)(C)CCO
This item is not returnable. View return policy
67-48-1
C5H14ClNO
139.62
MFCD00011721
SGMZJAMFUVOLNK-UHFFFAOYSA-M
choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
6209
CHEBI:133341
(2-hydroxyethyl)trimethylazanium chloride
[Cl-].C[N+](C)(C)CCO
Color White
Density 1.1g/cm3 at 20°C
Quantity 100 g
Formula Weight 139.6
Percent Purity ≥99%
Physical Form Hard Crystalline Powder
Chemical Name or Material Choline chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'