Läs mer
Choline Dihydrogen Citrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Choline Dihydrogen Citrate |
| CAS | 77-91-8 |
| Molekylformel | C6H7O7.C5H14NO |
| Kvantitet | 100 g |
| Synonym | 2-Hydroxy-1,2,3-propanetricarboxylic Acid Ion(1-) 2-Hydroxy-N,N,N-trimethylethanaminium, 2-Hydroxy-N,N,N-trimethylethanaminium Salt with 2-Hydroxy-1,2,3-propanetricarboxylic Acid, Choline Citrate, Citric acid, Ion(1-), Choline, 2-Hydroxyethyltrimethylammonium Dihydrogen Citrate, Cholex, Cholinvel, Chothyn, Cirrocolina, Citracholin, Citracholine, Citrocholine, Citrocol, Colicitrin, Delichol, Neurotropan |
| InChI formel | InChI=1S/C6H8O7.C5H14NO/c7-3(8)1-6(13,5(11)12)2-4(9)10;1-6(2,3)4-5-7/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);7H,4-5H2,1-3H3/q;+1/p-1 |
| LEDER | C[N+](C)(C)CCO.OC(=O)CC(O)(CC(=O)[O-])C(=O)O |
| IUPAC-namn | 3-carboxy-3,5-dihydroxy-5-oxo-pentanoate;2-hydroxyethyl(trimethyl)ammonium |
| Molekylvikt (g/mol) | 295.29 |
| Formel vikt | 295.1267 |