Läs mer
Chondroitin Sulfate, Bovine Trachea, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Chondroitin Sulfate |
| CAS | 9007-28-7 |
| Molekylformel | C13H21NO15S |
| Kvantitet | 250 mg |
| Synonym | Chondroitin, hydrogen sulfate, Chondroitinsulfuric acids (8CI), ChondroGuard, Chondroitin Q, Chondroitin polysulfate, Chondroitin sulfate, Chondroitin sulphate, Chondroitinsulfuric acid, Chonsurid, Cosamin DS, Marine Cartilage 40S, Marine Chondroitin, Mucosatum, Uracyst S 400 |
| InChI formel | InChI=1S/C13H21NO15S/c1-2(15)14-3-8(7(19)13(28-11(3)22)29-30(23,24)25)26-12-6(18)4(16)5(17)9(27-12)10(20)21/h3-9,11-13,16-19,22H,1H3,(H,14,15)(H,20,21)(H,23,24,25)/t3-,4+,5+,6-,7-,8-,9+,11-,12-,13-/m1/s1 |
| LEDER | CC(=O)N[C@@H]1[C@H]([C@H]([C@H](O[C@H]1O)OS(=O)(=O)O)O)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C(=O)O)O)O)O |
| IUPAC-namn | (2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Molekylvikt (g/mol) | 463.37 |