missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12126-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
CHRAC1 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.[supplied by OMIM, Apr 2004].Specifications
CHRAC1 | |
Polyclonal | |
Unconjugated | |
CHRAC1 | |
CHARC1, CHARC15, CHRAC 1, CHRAC 15, CHRAC1, CHRAC15, HuCHRAC15, YCL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315058, 54108, 93696 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JKP8, Q9NRG0 | |
Chrac1 | |
CHRAC1 Fusion Protein Ag2770 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |