Learn More
Macherey-Nagel™ Chromabond™ HR-XAW Polypropylene SPE Column
895.00 SEK - 2975.00 SEK
Specifications
Base Material | Spherical PS/DVB Copolymer |
---|---|
Column Type | SPE |
Particle Size | 85 μm |
Sorbent | Chromabond HR-XAW |
Type | Solid Phase Extraction Column |
Product Code | Brand | Column Capacity (Metric) | Pore Size | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity (Metric) | Pore Size | Description | Price | Quantity & Availability | |||
12757720
|
Macherey-Nagel™
730729 |
1 mL | 55 to 65 μm | Adsorbent Weight: 100mg |
1165.00 SEK
Pack of 30 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
12798192
|
Macherey-Nagel™
730728 |
1 mL | 55 to 65 Å | Adsorbent Weight: 30mg |
895.00 SEK
Pack of 30 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
12788212
|
Macherey-Nagel™
730748 |
3 mL | 55 to 65 Å | Adsorbent Weight: 200mg |
1435.00 SEK
Pack of 30 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
12748212
|
Macherey-Nagel™
730744 |
3 mL | 55 to 65 μm | Adsorbent Weight: 500mg |
2095.00 SEK
Pack of 30 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
12768212
|
Macherey-Nagel™
730747 |
3 mL | 55 to 65 μm | Adsorbent Weight: 60mg |
1280.00 SEK
Pack of 30 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
12777730
|
Macherey-Nagel™
730749 |
6 mL | 55 to 65 μm | Adsorbent Weight: 150mg |
2630.00 SEK
Pack of 30 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
12758212
|
Macherey-Nagel™
730745 |
6 mL | 55 to 65 μm | Adsorbent Weight: 500mg |
2975.00 SEK
Pack of 30 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||
Specifications
Spherical PS/DVB Copolymer | |
85 μm | |
Solid Phase Extraction Column |
SPE | |
Chromabond HR-XAW | |
850 m2/g |