Läs mer
Chromafenozide, MedChemExpress
Produktkod. 16493407
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16493407 50 mg
16407388 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16493407 Leverantör Medchem Express Leverantörsnummer HY17533
MedChemExpress Chromafenozide (ANS118) is a lepidopteran insecticide; it is highly effective in controlling various lepidopteran pests.
Chromafenozide (ANS118) is a lepidopteran insecticide
Specifikationer
Kemiskt namn eller material Chromafenozide
CAS 143807-66-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C24H30N2O3
Kvantitet 50 mg
Synonym ANS118
Löslighetsinformation DMSO : 250 mg/mL (633.70 mM; Need ultrasonic)
LEDER O=C(C1=CC=C2C(CCCO2)=C1C)NN(C(C3=CC(C)=CC(C)=C3)=O)C(C)(C)C
Molekylvikt (g/mol) 394.51
Formel vikt 394.51
Procent renhet 99.57%
Kvalitet Research
Fysisk form Solid