Learn More
Kimble Chase™ Chromaflex™ Glass Barrel
Brand:  Kimble Chase™ 420831-3010
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11813063
  • 3415.00 SEK / Each
Estimated Shipment: 09-07-2024
to see stock.
Specifications
25 mm
1 Each
Glass
Barrel
300 mm
25 mm