Learn More
Purolite™ Chromalite™ PCG Macroporous Hydrophobic Polymeric Adsorbent
Chromalite™ PCG resins are hydrophobic porous polydivinylbenzene adsorbent. Porosity and matrix are specific for purification by reversed-phase chromatography, solid phase extraction and adsorption of biomolecules including proteins, peptides, oligonucleotides and antibiotics.
Brand:  Purolite™ LS06725-94
Additional Details : CAS Number : 9003-70-7 Weight : 1.00000kg
 View more versions of this product
Product Code. 16824303
  • 17564.40 SEK / 1kg
Estimated Shipment: 01-12-2021
Add to basket
Description
Description
  • Reverse phase chromatography
  • Adsorption of biomolecules
  • Solid Phase Extraction
  • Highly hydrophobic
  • High chemical stability
  • More robust than silica materials
  • Easy column packing
  • Efficient regeneration
  • Compliant with FDA regulation
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifications
Specifications
White to off-white spherical beads
2°C to 20°C
50 to 100 μm
300 to 500 Å
1 kg
65 to 80%
PCG1200M
Macroporous Polydivinylbenzene Adsorbent
1 to 14 pH
>600 m2/g
Wet