Läs mer
RETSCH Chrome Steel Comfort Grinding Jars
Produktkod. 10789944 Handla allt Retsch Produkter
Change view
Kapacitet (metrisk):
50 ml
125 ml
250 ml
500 ml
Förpackningsstorlek:
Varje
Produktkod. Kapacitet (metrisk) unitSize
10789944 500 ml Varje
10402272 250 ml Varje
10044851 125 ml Varje
10339461 50 ml Varje
4 options
Produktkod. 10789944
Brand: RETSCH 01.462.0229
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Use these grinding jars with Retsch™ ball mills. Retsch™ Chrome Steel Comfort Grinding Jars feature an O-type sealing for safe operation.
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifikationer
Produkttyp Comfort Grinding Jar
Kapacitet (metrisk) 500 ml
För användning med (utrustning) For PM 100, PM 400
Material Chrome Steel